missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene 99.0+%, TCI America™

Catalog No. G022810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G022810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G022810G Supplier TCI America Supplier No. G022810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 489-84-9
Molecular Formula C15H18
Molecular Weight (g/mol) 198.309
MDL Number MFCD00003811
InChI Key FWKQNCXZGNBPFD-UHFFFAOYSA-N
Synonym guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
PubChem CID 3515
ChEBI CHEBI:5550
IUPAC Name 1,4-dimethyl-7-propan-2-ylazulene
SMILES CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

Specifications

Color Blue
Boiling Point 320°C
Quantity 10 g
Formula Weight 198.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Guaiazulene
EINECSNumber (9)-0413
RTECSNumber CO4790000
TSCA Yes