missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene, 98+%

Catalog No. AAA1769014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1769014 25 g
AAA1769006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1769014 Supplier Thermo Scientific Chemicals Supplier No. A1769014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 489-84-9 | C15H18 | 198.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is employed as antimicrobial agent and perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water, 0.1115 mg/L @ 25°C (est).

Notes
Store in dark. Protect from heat, oxidizing agents and heat. Keep it refrigerated.

Chemical Identifiers

CAS 489-84-9
Molecular Formula C15H18
Molecular Weight (g/mol) 198.309
MDL Number MFCD00003811
InChI Key FWKQNCXZGNBPFD-UHFFFAOYSA-N
Synonym guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
PubChem CID 3515
ChEBI CHEBI:5550
IUPAC Name 1,4-dimethyl-7-propan-2-ylazulene
SMILES CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

Specifications

Melting Point 31°C to 33°C
Density 0.976
Boiling Point 152°C to 154°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1365001
Sensitivity Light sensitive
Merck Index 14,4554
Solubility Information Soluble in alcohol,water,0.1115mg/L @ 25°C (est).
Formula Weight 198.31
Percent Purity ≥98%
Chemical Name or Material Guaiazulene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 207-701-2
RTECSNumber CO4790000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only