missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthers de glycérol de guaiacol 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 83°C |
| Color | Blanc |
| Boiling Point | 215°C |
| Quantity | 25 g |
| Formula Weight | 198.22 |
| Percent Purity | ≥98.0% (HPLC,E) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Guaiacol Glycerol Ether |
Sécurité et manipulation
| EINECSNumber | (3)-0576&(3)-0588 |
| RTECSNumber | TY8400000 |
| TSCA | Yes |