missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C5/TFIIIC63 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GTF3C5 is polypeptide 5 of general transcription factor IIIon Channel. Human transcription factor IIIon Channel (hTFIIIon Channel) is a multisubunit complex that directly recognizes promoter elements and recruits TFIIIB and RNA polymerase III.
Spécifications
Spécifications
| Antigène | GTF3C5/TFIIIC63 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | GTF3C5 |
| Numéro d’acquisition du gène | Q8R2T8, Q9Y5Q8 |
| Alias du gène | general transcription factor IIIC, polypeptide 5 (63kD), general transcription factor IIIC, polypeptide 5, 63kDa, TF3C-epsilon, TFIIIC 63 kDa subunit, TFiiiC2-63, TFIIIC63, TFIIICepsilon, transcription factor IIIC 63 kDa subunit, transcription factor IIIC subunit epsilon, transcription factor IIIC, 63 kD |
| Symboles de gène | Gtf3c5 |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |