Learn More
Cytiva GST Gene Fusion System pGEX Vectors
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
$1247.20 - $1501.26
Specifications
Quantity 25 μg
For Use With (Application) Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
Type pGEX Vector
View More Specs
Products 4
Catalog Number Mfr. No. Kit Contents Price Quantity  
Catalog Number Mfr. No. Kit Contents Price Quantity  
45-002-271
View Documents
Cytiva
28954549
4T-1
Each for $1,247.20
Only null left
45-002-277
View Documents
Cytiva
28954651
6P-3
Each for $1,418.62
Only null left
45-002-275
View Documents
Cytiva
28954648
6P-1
Each for $1,268.26
Only null left
45-002-282
View Documents
Cytiva
28954552
4T-3
Each for $1,501.26
Only null left
Description
Description
  • Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
  • Offers an internal laq1q gene for use in any E. coli host
  • With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
  • The vectors provide all three translational reading frames beginning with the EcoRI restriction site
  • Can use a wide variety of E. coli host strains cloning and expression
  • pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
  • pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [γ-32P]ATP using protein kinase
Specifications
Specifications
25 μg
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
pGEX Vector
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'