Learn More
Abcam Inc GSK503, EZH2 methyltransferase inhibitor, 5MG
Numéro de catalogue. 502683344
Encompass
MW 526.7 Da. Potent (IC = nM; Ki = 3 nM) and specific EZH2 methyltransferase inhibitor. Selective against EZH1 (IC = 633 nM). At least 4000-fold selective over a panel of 20 other human methyltransferases. Highly selective against a range of human histone deacetylases, GPCRs, kinases and ion channels. Reduces cellular H3K27me3 levels. Inhibits growth of diffuse large B-cell lymphoma cells. Inhinits melanoma growth and metastasis in mouse models.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502683344 Fournisseur Abcam Inc Code fournisseur AB2698195MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 526.7 Da. Potent (IC = nM; Ki = 3 nM) and specific EZH2 methyltransferase inhibitor. Selective against EZH1 (IC = 633 nM). At least 4000-fold selective over a panel of 20 other human methyltransferases. Highly selective against a range of human histone deacetylases, GPCRs, kinases and ion channels. Reduces cellular H3K27me3 levels. Inhibits growth of diffuse large B-cell lymphoma cells. Inhinits melanoma growth and metastasis in mouse models.
The product is subject to the following: Abcam Restricted Use Statement