Learn More
GSC Go Science Crazy Mini Motor Model
Catalog No. S13684
Encompass
Change view
Click to view available options
Motor, mini motor, model
Catalog No.
S13684 Motor, mini motor, model
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13684 Supplier GSC Go Science Crazy Supplier No. 418470
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Not a toy. Product intended for ages 13 and up.
A small AC/DC motor that helps students understand permanent and electromagnet motors. The GSC Mini Motor Model is a model of an AC/DC motor that demonstrates permanent and electromagnet motors. All the parts are packed in a protective plastic case that is used as the base of the motor during assembly. Everything is included.
  • Visually show how a motor works
  • AC/DC current
  • Instructions included
Specifications
Electrical Requirements Type: Direct Current (DC) Alternating Current (AC)
Width (English) 6 in.
Height (English) 4 in.