missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Growth Hormone Releasing Factor (GRF) (1-29) amide, human

Catalog No. AAJ61111EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61111EXD 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61111EXD Supplier Thermo Scientific Chemicals Supplier No. J61111EXD

A peptide hormone that stimulates release of the growth hormone

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86168-78-7

Specifications

Content And Storage Ambient temperatures
Format Powder
Quantity 0.5 mg

RUO – Research Use Only