missing translation for 'onlineSavingsMsg'
Learn More

Griseofulvin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012168
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012168 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012168 Supplier Merck Emd Millipore Supplier No. PHR15341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 126-07-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H17 ClO6
MDL Number MFCD00082343
Synonym Griseofulvin; (2 S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3 H],1′-[2 ]cyclohexene)-3,4′-dione
Type Certified Reference Material
Quantity 1 g
Formula Weight 352.77
Grade Certified Reference Material