missing translation for 'onlineSavingsMsg'
Learn More

(+)-Griseofulvin, 97%

Catalog No. AAB2352122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2352122 100 g
AAB2352106 5 g
AAB2352114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2352122 Supplier Thermo Scientific Chemicals Supplier No. B2352122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126-07-8 | C17H17ClO6 | 352.767 g/mol

Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 126-07-8
Molecular Formula C17H17ClO6
Molecular Weight (g/mol) 352.767
MDL Number MFCD00082343
InChI Key DDUHZTYCFQRHIY-RBHXEPJQSA-N
Synonym griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine
PubChem CID 441140
ChEBI CHEBI:27779
IUPAC Name (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
SMILES CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC

Specifications

Melting Point 218°C to 220°C
Odor Odorless
Quantity 100 g
Beilstein 95226
Merck Index 14,4549
Solubility Information Soluble in dimethyl formamide,ethanol,methanol,chloroform,DMSO,tetrachloroethane. Insoluble in water.
Formula Weight 352.77
Percent Purity 97%
Chemical Name or Material (+)-Griseofulvin
Hazard Category H317-H351-H360FD
Hazard Statement GHS H Statement
H360-H351-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 204-767-4
RTECSNumber WG9800000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only