Learn More
Greiner Bio-One VACUETTE™ K2EDTA Blood Collection Tubes
Catalog No. 22040161
Change view
Click to view available options
Capacity (Metric):
2 mL
6 mL
Closure Color:
Lavender with Black Ring
Lavender with White Ring
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22040161 6 mL Lavender with Black Ring
22040307 2 mL Lavender with White Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22040161 Supplier Greiner Bio-One Supplier No. 456002
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K2EDTA tubes contain dipotassium K2EDTA which blocks the coagulation cascade.
  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Made of virtually unbreakable PET plastic
  • Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Primarily in hematology
  • Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
  • K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
  • Pink caps for immunohematology are also available
  • CE marked
Specifications
Product Type K2 EDTA Tube
Additive K2 EDTA
Material PET
Sterility Sterile
Closure Type Safety Pull Cap
Capacity (Metric) 6 mL
Closure Color Lavender with Black Ring
For Use With (Application) Blood Collection