Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner
Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
$401.75 - $16,273.19
Specifications
| Color | Green |
|---|---|
| Material | Thermoset |
| Sterility | Non-sterile |
| Thread Size (Metric) | 72 mm |
| Liner Material | F217, PTFE |
| Catalog Number | Mfr. No. | Neck Finish | Product Type | Packaging | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Product Type | Packaging | Price | Quantity & Availability | |||
|
02913105
|
Qorpak™
272499 |
8-425 | Lined Cap (Ribbed) | Bulk |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
02913106
|
Qorpak™
272635 |
8-425 | Lined Cap (Ribbed) | Bagged |
Case of 500 Each for $554.50
|
|
|||
|
02913107
|
Qorpak™
272632 |
13-425 | Lined Cap (Ribbed) | Bagged |
Case of 500 Each for $401.75
|
|
|||
|
02913108
|
Qorpak™
272490 |
15-425 | Lined Cap (Ribbed) | Bulk |
Case of 12600 Each for $7,275.69
|
|
|||
|
02913109
|
Qorpak™
272633 |
15-425 | Lined Cap (Ribbed) | Bagged |
Case of 500 Each for $482.63
|
|
|||
|
02913110
|
Qorpak™
272491 |
18-400 | Lined Cap (Ribbed) | Bulk |
Case of 7600 Each for $6,758.79
|
|
|||
|
02913111
|
Qorpak™
272634 |
18-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $613.09
|
|
|||
|
02913112
|
Qorpak™
272492 |
20-400 | Lined Cap (Smooth) | Bulk |
Case of 6300 Each for $5,780.53
|
|
|||
|
02913113
|
Qorpak™
272626 |
20-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $700.63
|
|
|||
|
02913114
|
Qorpak™
272493 |
22-400 | Lined Cap (Smooth) | Bulk |
Case of 5300 Each for $5,564.38
|
|
|||
|
02913115
|
Qorpak™
272627 |
22-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $833.00
|
|
|||
|
02913116
|
Qorpak™
272494 |
24-400 | Lined Cap (Smooth) | Bulk |
Case of 4400 Each for $5,312.41
|
|
|||
|
02913117
|
Qorpak™
272628 |
24-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $927.21
|
|
|||
|
02913118
|
Qorpak™
272495 |
28-400 | Lined Cap (Smooth) | Bulk |
Case of 3200 Each for $4,703.47
|
|
|||
|
02913119
|
Qorpak™
272629 |
28-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $1,139.66
|
|
|||
|
02913120
|
Qorpak™
272496 |
33-400 | Lined Cap (Smooth) | Bulk |
Case of 2300 Each for $4,346.53
|
|
|||
|
02913121
|
Qorpak™
272630 |
33-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $1,435.35
|
|
|||
|
02913122
|
Qorpak™
272497 |
38-400 | Lined Cap (Smooth) | Bulk |
Case of 1680 Each for $4,150.12
|
|
|||
|
02913123
|
Qorpak™
272631 |
38-400 | Lined Cap (Smooth) | Bagged |
Case of 500 Each for $1,911.47
|
|
|||
Description
- The color green easily identifies PTFE liner system
- F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
- Provides odor and impact resilience for a tight seal with a low moisture transmission rate
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
- Available in smooth and ribbed style
Specifications
| Green | |
| Non-sterile | |
| F217, PTFE | |
| Not Autoclavable |
| Thermoset | |
| 72 mm | |
| Reusable |