Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner
Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Supplier:  Qorpak™ 272634
 View more versions of this product
Catalog No. 02913111
Only null left
Description
Description
  • The color green easily identifies PTFE liner system
  • F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
  • Available in smooth and ribbed style
Specifications
Chemical Identifiers
Green
Non-sterile
F217, PTFE
Not Autoclavable
Thermoset
72 mm
Reusable
Specifications
Green
500
Non-sterile
F217, PTFE
Reusable
Not Autoclavable
Thermoset
Bagged
72 mm
18-400
Lined Cap (Smooth)
Product Suggestions
Product Suggestions