Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner
Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Supplier: Qorpak™ 272494
Description
- The color green easily identifies PTFE liner system
- F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
- Provides odor and impact resilience for a tight seal with a low moisture transmission rate
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
- Available in smooth and ribbed style
Chemical Identifiers
| Green | |
| Non-sterile | |
| F217, PTFE | |
| Not Autoclavable |
| Thermoset | |
| 72 mm | |
| Reusable |
Specifications
| Green | |
| 4400 | |
| Non-sterile | |
| F217, PTFE | |
| Reusable | |
| Not Autoclavable |
| Thermoset | |
| Bulk | |
| 72 mm | |
| 24-400 | |
| Lined Cap (Smooth) |