Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner
Catalog No. 02913117
Change view
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
8-425
Product Type:
Lined Cap (Ribbed)
Lined Cap (Smooth)
Packaging:
Bagged
Bulk
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type Packaging
02913117 24-400 Lined Cap (Smooth) Bagged
02913105 8-425 Lined Cap (Ribbed) Bulk
02913107 13-425 Lined Cap (Ribbed) Bagged
02913108 15-425 Lined Cap (Ribbed) Bulk
02913109 15-425 Lined Cap (Ribbed) Bagged
02913110 18-400 Lined Cap (Ribbed) Bulk
02913111 18-400 Lined Cap (Smooth) Bagged
02913112 20-400 Lined Cap (Smooth) Bulk
02913114 22-400 Lined Cap (Smooth) Bulk
02913115 22-400 Lined Cap (Smooth) Bagged
02913116 24-400 Lined Cap (Smooth) Bulk
02913118 28-400 Lined Cap (Smooth) Bulk
02913119 28-400 Lined Cap (Smooth) Bagged
02913120 33-400 Lined Cap (Smooth) Bulk
02913121 33-400 Lined Cap (Smooth) Bagged
02913122 38-400 Lined Cap (Smooth) Bulk
02913123 38-400 Lined Cap (Smooth) Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02913117 Supplier Qorpak™ Supplier No. 272628
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
  • The color green easily identifies PTFE liner system
  • F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
  • Available in smooth and ribbed style
Specifications
Color Green
Material Thermoset
No. per Case 500
Packaging Bagged
Sterility Non-sterile
Thread Size (Metric) 72 mm
Liner Material F217, PTFE
Neck Finish 24-400
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Lined Cap (Smooth)
Show More Show Less