missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB10 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GRB7, a SH2 domain protein, has a single SH2 domain at its C-terminal, a central region with similarity to Ras GAP, and a proline-rich N terminus. A related SH2 domain-containing protein, GRB10, exhibits a high degree of homology with GRB7. GRB10 undergoes serine but not tyrosine phosphorylation in response to EGF treatment, but appears to bind to the EGF receptor poorly. GRB10 maps to mouse chromosome 11, in close proximity to the EGF receptor. Similarly, GRB7 maps to the same mouse chromosome near the EGF receptor-related protein HER2.
Spécifications
Spécifications
| Antigène | GRB10 |
| Applications | Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugué | Unconjugated |
| Formule | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gène | GRB10 |
| Numéro d’acquisition du gène | Q13322 |
| Alias du gène | GRB10 adapter protein; GRB10 adaptor protein; OTTHUMP00000197988; OTTHUMP00000197989; OTTHUMP00000197990; OTTHUMP00000197991; OTTHUMP00000197993; OTTHUMP00000197994; insulin receptor-binding protein Grb-IR; maternally expressed gene 1 |
| Symboles de gène | GRB10 |
| Afficher plus de résultats |