missing translation for 'onlineSavingsMsg'
Learn More

Graphite plate, resin impregnated, 6.35mm (0.25in) thick, Thermo Scientific Chemicals

Catalog No. AA41831GH
Change view
Click to view available options
Quantity:
100 x 100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41831GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41831GH Supplier Thermo Scientific Chemicals Supplier No. 041831GH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Graphite is basically used for casting and polishing purposes, moulding & carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy. It is mostly used for evaporating carbon onto specimens being prepared for either x-ray microanalysis (EDS or WDS) or transmission electron microscopy (TEM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Graphite plate
Name Note Resin impregnated, 6.35mm (0.25 in.) thick
MDL Number MFCD00144065
Quantity 100 x 100 mm
Solubility Information Insoluble in water.

RUO – Research Use Only