Learn More
Thermo Scientific Chemicals Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Description | Unpurified extruded graphite |
| Material | Graphite |
| For Use With (Application) | Metallurgy |
| Quantity | 50 Pieces |
| Capacity (Metric) | 7.5 cc |
| Product Type | Graphite fusion crucible |
| Shape | Drillpoint |
RUO – Research Use Only