missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals

Catalog No. AA43083RX
Change view
Click to view available options
Quantity:
30 x 30 cm
30 x 100 cm
30 x 500 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43083RX 30 x 500 cm
AA43083RF 30 x 30 cm
AA43083CJ 30 x 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AA43083RX Supplier Thermo Scientific Chemicals Supplier No. 043083RX
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00144065

Specifications

Quantity 30 x 500 cm
Percent Purity 97%
Physical Form Foil
Assay (metals basis)
Chemical Name or Material Graphite foil, 1mm (0.04 in.) thick
EINECSNumber 231-955-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only