missing translation for 'onlineSavingsMsg'
Learn More

Graphite, colloidal, lubricant, aerosol spray, Thermo Scientific Chemicals

Catalog No. AA417751X
Change view
Click to view available options
Quantity:
10 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA417751X 10 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA417751X Supplier Thermo Scientific Chemicals Supplier No. 0417751X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

Dry film lubricant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Quantity 10 oz.
Chemical Name or Material Graphite
Hazard Category H225-H304-H315-H319-H335-H336-H361d
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 2.1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only