missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 0.50mm

Numéro de catalogue. p-4865466
Change view
Click to view available options
Quantity:
5 Pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA42073KJ 5 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AA42073KJ Fournisseur Thermo Scientific™ Code fournisseur 42073KJ
Il en reste null

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Product Type Ruby Sphere
Diameter (Metric) 0.50 mm
Grade Grade 25
Quantity 5 Pieces