missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR83 Monoclonal Antibody (8A6D3)
GREENER_CHOICE

Numéro de catalogue. FERMA548706
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
FERMA548706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. FERMA548706 Fournisseur Invitrogen™ Code fournisseur MA548706

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

GPR83 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.

Spécifications

Antigène GPR83
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8A6D3
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.05% sodium azide
Gène GPR83
Numéro d’acquisition du gène P30731, Q9NYM4
Alias du gène AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82
Symboles de gène GPR83
Espèces hôtes Mouse
Immunogène Purified recombinant fragment of human GPR83 (AA: extra mix) expressed in E. Coli.
Méthode de purification Protein G
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 10888, 14608
Espèces cibles Human, Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats