missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR108 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533604
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Mouse, Rat, Hamster, Bovine (100%) Gibbon, Marmoset, Panda, Dog (93%) Pufferfish (87%).
GPR108 is a protein coding gene.Identifiants chimiques
| GPR108 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat, Bovine, Hamster | |
| GPR108 | |
| 1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108 | |
| GPR108 | |
| Primary | |
| Antigen affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.1% sodium azide | |
| Q148L1, Q6P6V6, Q91WD0, Q9NPR9 | |
| 100764254, 316136, 507830, 56927, 78308 | |
| Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human GPR108. | |
| Antibody |
Spécifications
| GPR108 | |
| Polyclonal | |
| Unconjugated | |
| GPR108 | |
| 1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100764254, 316136, 507830, 56927, 78308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q148L1, Q6P6V6, Q91WD0, Q9NPR9 | |
| GPR108 | |
| Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human GPR108. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Hamster | |
| Antibody | |
| IgG |