missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Polyethylene Transfer Pipets

Catalog No. p-4429524
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 mL
3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07201926 1 mL
07201927 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 07201926 missing translation for 'mfr' Corning™ missing translation for 'supplierNo' PAS1A01
Only null left

With metric graduations

  • Polyethylene (PE)
  • Complete and quick removal of liquid
  • Peelable bag

Specifications

Material Polyethylene
Sterility Sterile
Graduated Yes
Quantity 2000/Cs.
Packaging Type Peelable bag
Volume (Metric) 1 mL
Length (Metric) 15 cm
Type Transfer Pipet
Disposable Single-use