missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Blender Bag Clips

Description
GosselinTM clip for blender bags are ideal for closing 80 mL and 400 mL blender bags between the sample preparation steps.
Specifications
Specifications
| Type | Clip |
| Material | Polyethylene |
| For Use With (Equipment) | Blender Bag |
| Color | Natural |