missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Gold Vacuum Pump Oil, Corrosion Resistant

Catalog No. 16985649
Change view
Click to view available options
Format:
Oil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
16985649 Oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16985649 Supplier Labconco™ Supplier No. 7772700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gold Vacuum Pump Oil is a double distilled synthetic hydrocarbon oil optimized for excellent chemical resistance.

  • No aromatic compounds or sulfur that create varnishing, sludging, and carbon build up when pumps are used with corrosives.

Specifications

Product Type Rotary-vane Pump Oil
Format Oil
For Use With (Application) Laboratory Vacuum Pumps
For Use With (Equipment) Welch Rotary Vane Vacuum Pumps
Volume (English) 33.8 oz.
Volume (Metric) 1 L