missing translation for 'onlineSavingsMsg'
Learn More

Gold AA Standard, Certified, 1000 ppm (1mL = 1mg Au), LabChem™

Catalog No. p-5388655
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC148907 125 mL Amber Glass
LC148901 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC148907 Supplier LabChem Supplier No. LC148907
Only null left

Chemical Identifiers

CAS 16961-25-4 , 7647-01-0 , 7732-18-5
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Packaging Amber Glass
CAS Max % 89.82, 9.98, 0.2
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm (1mL = 1mg Au)
Linear Formula HAuCl4·3H2O
UN Number UN3264
Color Yellow
Solubility Soluble in water
Quantity 125 mL
Formula Weight 393.83
Grade Certified
Chemical Name or Material Gold AA Standard, 1000ppm (1mL = 1mg Au)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature