missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Goat Serum

Catalog No. MP92939154
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP92939154 500 mL
MP92939149 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP92939154 Supplier MP Biomedicals™ Supplier No. 092939154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.

This serum is produced from whole blood collected from healthy adult animals.

Specifications

Color Reddish brown
Content And Storage Store at -20°C
Form Liquid
Quantity 500 mL
Sterilization Method 0.2 μm filtration
Source Whole blood collected from healthy adult animals
For Use With (Application) Cell Culture
pH 7 to 8