missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgG2a Secondary Antibody
Description
PA1-28885 detects Mouse IgG2a from mouse samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Mouse IgG2a |
| Applications | ELISA, Immunodiffusion, Immunohistochemistry, Lateral Flow, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.1% sodium azide; pH 7.2 |
| Espèces hôtes | Goat |
| Immunogène | Highly purified mouse IgG2a in FreundOs adjuvant |
| Méthode de purification | Antigen affinity chromatography |
| Quantité | 500 μg |
| Afficher plus de résultats |