missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, HRP

Numéro de catalogue. H10307
Modifier l'affichage
Click to view available options
Quantité:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
H10307 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H10307 Fournisseur Invitrogen™ Code fournisseur H10307

Goat Polyclonal Secondary Antibody

Goat anti-Human IgG (H+L) Secondary Antibody for ELISA, Western Blot

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Human IgG (H+L)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugué HRP
Formule PBS with 40% glycerol, 4mg/mL BSA and no preservative
Espèces hôtes Goat
Méthode de purification Purified
Quantité 2 mL
Statut réglementaire ASR
Primaire ou secondaire Secondary
Espèces cibles Human
Contenu et stockage 4°C
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats