missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig λ chain Goat anti-Human, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Goat anti-Human Ig λ chain Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA for the detection of Human Ig λ chain.
Spécifications
Spécifications
| Antigène | Goat anti-Human Ig λ chain |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugué | Alkaline Phosphatase (AP) |
| Formule | Purified by affinity chromatography on pooled human Igs with lambda light chains covalently linked to agarose. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative. |
| Espèces hôtes | Goat |
| Immunogène | pooled antisera from goats hyperimmunized with human myeloma proteins with lambda light chains. |
| Méthode de purification | Affinity Purified |
| Quantité | 1 mL |
| Primaire ou secondaire | Secondary |
| Afficher plus de résultats |
For Research Use Only