missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, TRITC
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigen | Chicken IgY (H+L) Cross-Adsorbed |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | TRITC |
| Formulation | 0.02M potassium phosphate with 10mg/mL BSA, 0.15M NaCl and 0.01% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Full length native chicken IgY (purified). |
| Purification Method | IgG fraction |
| Quantity | 100 μg |
| Afficher plus de résultats |