missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals GNF-5, 95%

Catalog No. AC468191000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468191000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468191000 Supplier Thermo Scientific Chemicals Supplier No. 468191000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 778277-15-9
Molecular Formula C20H17F3N4O3

Specifications

Assay Percent Range 95%
Quantity 100 mg
Formula Weight 418.37
Chemical Name or Material GNF-5
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

May cause an allergic skin reaction.

Causes serious eye irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Gently wash with plenty of soap and water.

Call a POISON CENTER or doctor/physician.

If eye irritation persists: Get medical advice/attention.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only