CAS RN 778277-15-9
IUPAC Name:
N-(2-hydroxyethyl)-3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Synonyms:
N-(2-hydroxyethyl)-3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Molecular Weight (g/mol):
418.38
Molecular Formula:
C20H17F3N4O3
InChi Key:
IIQUYGWWHIHOCF-UHFFFAOYSA-N
SMILES:
OCCNC(=O)C1=CC=CC(=C1)C1=CC(NC2=CC=C(OC(F)(F)F)C=C2)=NC=N1
1
–
2
of
2
results
1
Thermo Scientific Chemicals GNF-5, 95%
CAS: 778277-15-9 Molecular Formula: C20H17F3N4O3
2
R&D Systems™ GNF 5
CAS: 778277-15-9 Molecular Formula: C20H17F3N4O3