missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GNF 5

Catalog No. 490850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
490850 50 mg
490810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 490850 Supplier Tocris Bioscience™ Supplier No. 4908/50
Only null left

Selective allosteric inhibitor of Bcr-Abl; analog of GNF 2 (Cat. No. 4399)

Selective, non-ATP competitive allosteric inhibitor of Bcr-Abl (IC50 = 220 nM for wild-type Abl). Binds the myristate-binding site of Abl. Acts in combination with nilotinib to inhibit T315I Bcr-Abl in vitro and in vivo. Analog of GNF 2 (Cat. No. 4399).

Chemical Identifiers

CAS 778277-15-9
Molecular Formula C20H17F3N4O3

Specifications

Quantity 50 mg
Formula Weight 418.37
Molecular Weight (g/mol) 418.376
Chemical Name or Material GNF 5
Recommended Storage Store at +4°C