missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPR2 Monoclonal Antibody (K49018_14G3), Invitrogen™
Mouse Monoclonal Antibody
$1,601.06 - $2,821.92
Specifications
| Antigen | GMPR2 |
|---|---|
| Clone | K49018_14G3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: MPHIDNDVKL DFKDVLLRPK RSTLKSRSEV DLTRSFSFRN SKQTYSGVPI IAANMDTVGT FEMAKVLCKF SLFTAVHKHY SLVQWQEFAG QNPDCLEHLA ASSGTGSSDF EQLEQILEAI PQVKYICLDV ANGYSEHFVE FVKDVRKRFP QHTIMAGNVV TGEMVEELIL SGADIIKVGI GPGSVCTTRK KTGVGYPQLS AVMECADAAH GLKGHIISDG GCSCPGDVAK AFGAGADFVM LGGMLAGHSE SGGELIERDG KKYKLFYGMS SEMAMKKYAG GVAEYRASEG KTVEVPFKGD VEHTIRDILG GIRSTCTYVG AAKLKELSRR TTFIRVTQQV NPIFSEAC
GMPR2 Monoclonal antibody specifically detects GMPR2 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| GMPR2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog | |
| Recombinant human GMPR2 (immunogen range is 1-348). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GMPR2 |
| K49018_14G3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9P2T1 | |
| 51292 | |
| IgG1 | |
| Protein A | |
| Antibody |