missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GMPR2 Monoclonal Antibody (K49018_14G3), Invitrogen™

Catalog No. MA556150
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556150 200 μL
MA555770 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556150 Supplier Invitrogen™ Supplier No. MA556150
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MPHIDNDVKL DFKDVLLRPK RSTLKSRSEV DLTRSFSFRN SKQTYSGVPI IAANMDTVGT FEMAKVLCKF SLFTAVHKHY SLVQWQEFAG QNPDCLEHLA ASSGTGSSDF EQLEQILEAI PQVKYICLDV ANGYSEHFVE FVKDVRKRFP QHTIMAGNVV TGEMVEELIL SGADIIKVGI GPGSVCTTRK KTGVGYPQLS AVMECADAAH GLKGHIISDG GCSCPGDVAK AFGAGADFVM LGGMLAGHSE SGGELIERDG KKYKLFYGMS SEMAMKKYAG GVAEYRASEG KTVEVPFKGD VEHTIRDILG GIRSTCTYVG AAKLKELSRR TTFIRVTQQV NPIFSEAC

Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).

Specifications

Antigen GMPR2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K49018_14G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene GMPR2
Gene Accession No. Q9P2T1
Gene Alias 1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog
Gene Symbols GMPR2
Host Species Mouse
Immunogen Recombinant human GMPR2 (immunogen range is 1-348).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51292
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less