missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPR2 Monoclonal Antibody (K49018_14G3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555770
Description
Sequence of this protein is as follows: MPHIDNDVKL DFKDVLLRPK RSTLKSRSEV DLTRSFSFRN SKQTYSGVPI IAANMDTVGT FEMAKVLCKF SLFTAVHKHY SLVQWQEFAG QNPDCLEHLA ASSGTGSSDF EQLEQILEAI PQVKYICLDV ANGYSEHFVE FVKDVRKRFP QHTIMAGNVV TGEMVEELIL SGADIIKVGI GPGSVCTTRK KTGVGYPQLS AVMECADAAH GLKGHIISDG GCSCPGDVAK AFGAGADFVM LGGMLAGHSE SGGELIERDG KKYKLFYGMS SEMAMKKYAG GVAEYRASEG KTVEVPFKGD VEHTIRDILG GIRSTCTYVG AAKLKELSRR TTFIRVTQQV NPIFSEAC
Catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides (PubMed:12009299, PubMed:12669231, PubMed:16359702, PubMed:22037469). Plays a role in modulating cellular differentiation (PubMed:12669231).Chemical Identifiers
| GMPR2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog | |
| Recombinant human GMPR2 (immunogen range is 1-348). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GMPR2 |
| K49018_14G3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9P2T1 | |
| 51292 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| GMPR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9P2T1 | |
| GMPR2 | |
| Recombinant human GMPR2 (immunogen range is 1-348). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K49018_14G3 | |
| Unconjugated | |
| GMPR2 | |
| 1810008P16Rik; 5730544D12Rik; AA959850; GMP reductase 2; GMPR 2; GMPR2; Guanosine 5'-monophosphate oxidoreductase 2; guanosine monophosphate reductase 2; guanosine monophosphate reductase isolog | |
| Mouse | |
| Protein A | |
| RUO | |
| 51292 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |