missing translation for 'onlineSavingsMsg'
Learn More

Compass Biomedical™ GMP PLUS™ Human Platelet Lysate

Numéro de catalogue. PLSGB1000
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PLSGB1000 1000 mL
PLSGB0100 100 mL
PLSGB0250 250 mL
PLSGB0500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. PLSGB1000 Fournisseur Compass Biomedical™ Code fournisseur PLSGB1000

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Advanced human platelet lysate that is safe and consistent for the rapid expansion of human mesenchymal cell cultures.

  • Potent: Human derived product rich in growth factors, cytokine, and other proteins
  • Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.

Spécifications

For Use With (Application) Cell Culture, Stem Cell Culture
Quantity 1000 mL
Purity GMP Grade
Host Species Human
Product Type Human Platelet Lysate
Content And Storage Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended.
Format Bottle
Research Category GMP