missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Compass Biomedical™ GMP PLUS™ Cryobag Human Platelet Lysate
Description
- Potent: Human derived product rich in growth factors, cytokine, and other proteins
- Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.
Spécifications
Spécifications
| For Use With (Application) | Cell Culture, Stem Cell Culture |
| Quantity | 250 mL |
| Purity | GMP Grade |
| Host Species | Human |
| Product Type | Human Platelet Lysate |
| Content And Storage | Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended. |
| Format | Cryobag |
| Research Category | GMP |