missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid, Technical, 70-72%, Spectrum™ Chemical

Catalog No. 18603032
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603032 500 mL Amber Glass Bottle
18603033 4 L Amber Glass Bottle
18603031 20 L Poly Drum
18603034 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603032 Supplier Spectrum Chemical Mfg Cor Supplier No. GL185500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

GL185, C2H4O3

Spectrum™ Chemical Glycolic Acid, 70 Percent, Technical , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.

Chemical Identifiers

CAS 79-14-1 , 7732-18-5
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.05
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
IUPAC Name 2-hydroxyacetic acid
SMILES OCC(O)=O

Specifications

CAS Max % 70%, 30%
UN Number UN3265
Percent Purity 70 to 72%
Grade Technical
Quantity 500 mL
Packaging Amber Glass Bottle