missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid (ca. 70% in Water, ca. 12mol/L), TCI America™

Catalog No. G011025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G011025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G011025G Supplier TCI America Supplier No. G011025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-14-1
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00004312
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

Melting Point 10°C
Boiling Point 112°C
Color Yellow
Quantity 25 g
Physical Form Liquid
UN Number 3265
Formula Weight 76.05
Chemical Name or Material Glycolic Acid (ca. 70% in Water, ca. 12mol/L)
EINECSNumber (2)-1346
RTECSNumber MC5250000
TSCA Yes