missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Ultrapure Bioreagent, J.T. Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$195.29 - $1,315.82

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
02002943
View Documents Promotion Details
Avantor J.T.Baker
405900
250 g Poly Bottle
Each for $195.29
Only null left
 
02002942
View Documents Promotion Details
Avantor J.T.Baker
405902
1 kg Poly Bottle
Each for $348.26
Only null left
 
02002947
View Documents Promotion Details
Avantor J.T.Baker
405906
5 kg Poly Pail
Each for $1,315.82
Only null left
 
Description

Description

    For Electrophoresis, Liquid Chromatography and Molecular Biology Applications; For Laboratory,Research,or Manufacturing Use
Specifications

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O

Specifications

56-40-6
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
750
75.07
1L = 1.16kg
C2H5NO2
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
75.07
CHEBI:15428
Ultrapure Bioreagent
Glycine
Videos
SDS
Documents

Documents