missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02002943
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002943 250 g Poly Bottle
02002942 1 kg Poly Bottle
02002947 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02002943 Supplier Avantor Supplier No. 405900

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis, Liquid Chromatography and Molecular Biology Applications

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Packaging Poly Bottle
Quantity 250 g
Formula Weight 75.07
Grade Ultrapure Bioreagent
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine