missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-butyl ester, 97%

Catalog No. AAL1625806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1625806 5 g
AAL1625803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1625806 Supplier Thermo Scientific Chemicals Supplier No. L1625806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6456-74-2 | C6H13NO2 | 131.175 g/mol

Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine tert-butyl ester is used in the preparation of Schiff base by reacting with benzophenone. The resultant Schiff base undergoes asymmetric alkylation with arylmethyl bromides in the presence of O-allyl-N-(9-anthracenylmethyl)cinchonidinium bromide as chiral phase transfer catalyst to give guanidine-containing pentacyclic compound.

Solubility
Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6456-74-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00038194
InChI Key SJMDMGHPMLKLHQ-UHFFFAOYSA-N
Synonym tert-butyl glycinate, glycine tert-butyl ester, glycine, 1,1-dimethylethyl ester, tert-butyl aminoacetate, h-gly-otbu, t-butyl glycinate, glycin-tert-butyl ester, 1,1-dimethylethyl glycinate, glycine t-butyl ester, aminoacetic acid tert-butyl ester
PubChem CID 151417
IUPAC Name tert-butyl 2-aminoacetate
SMILES CC(C)(C)OC(=O)CN

Specifications

Boiling Point 29°C to 31°C (2mmHg)
Refractive Index 1.424
Beilstein 2039000
Solubility Information Miscible with acetone and methanol. Slightly miscible with ehtanol. Immiscible with water.
Density 0.96
Quantity 5 g
Formula Weight 131.18
Percent Purity 97%
Sensitivity Air sensitive
Chemical Name or Material Glycine tert-butyl ester
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only