missing translation for 'onlineSavingsMsg'
Learn More

Glycine Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014638
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014638 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014638 Supplier Merck Emd Millipore Supplier No. PHR11111G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 56-40-6
Linear Formula NH2CH2COOH
MDL Number MFCD00008131
Melting Point 240°C (decomposition)
Synonym Aminoacetic acid; Aminoethanoic acid; Glycocoll
Quantity 1 g
Formula Weight 75.07
Grade Certified Reference Material