missing translation for 'onlineSavingsMsg'
Learn More

Glycine, 10% (w/v), Ricca Chemical

Catalog No. p-3251724
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
30 gal.
Packaging:
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
32834 120 mL Natural Poly Bottle
328316 500 mL Natural Poly Bottle
328332 1 L Natural Poly Bottle
328330 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 32834 Supplier Ricca Chemical Company Supplier No. 32834
Only null left

Chemical Identifiers

CAS 56-40-6 , 7732-18-5
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 4
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Glycine
Show More Show Less