missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.0

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

$160.29 - $236.55

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62527AK
View Documents
Thermo Scientific Chemicals
J62527AK
250 mL
Each for $160.29
Only null left
 
AAJ62527AP
View Documents
Thermo Scientific Chemicals
J62527AP
500 mL
Each for $236.55
Only null left
 
Description

Description

Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Not miscible or difficult to mix in water. Slightly soluble in acetone. Insoluble in alcohol, and ether.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
NCC(O)=O
C2H5NO2
MFCD00008131
2-aminoacetic acid

Specifications

56-40-6
3.0
250 mL
Odorless
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
Glycine
0.2 M
Liquid
C2H5NO2
MFCD00008131
NCC(O)=O
75.07
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only