missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.0

Catalog No. AAJ62527AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62527AK 250 mL
AAJ62527AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62527AK Supplier Thermo Scientific Chemicals Supplier No. J62527AK

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Not miscible or difficult to mix in water. Slightly soluble in acetone. Insoluble in alcohol, and ether.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Concentration 0.2 M
pH 3.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material Glycine
Recommended Storage Ambient temperatures

RUO – Research Use Only