missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Monostearate, Powder, Food Grade, Spectrum™ Chemical
GL149, 31566-31-1, C21H42O4
$342.60 - $7642.71
Chemical Identifiers
| CAS | 31566-31-1 |
|---|---|
| Molecular Formula | C21H42O4 |
| Molecular Weight (g/mol) | 358.56 |
| InChI Key | VBICKXHEKHSIBG-UHFFFAOYNA-N |
| IUPAC Name | 2,3-dihydroxypropyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603000
|
Spectrum Chemical Mfg Cor
GL149500GM |
500 g | Poly Bottle |
Each for $342.60
|
|
||||
|
18603001
|
Spectrum Chemical Mfg Cor
GL14925KG |
2.5 kg | Poly Pail |
Each for $1,070.14
|
|
||||
|
18603002
|
Spectrum Chemical Mfg Cor
GL14912KG |
12 kg | Poly Pail |
Each for $2,757.51
|
|
||||
|
18603003
|
Spectrum Chemical Mfg Cor
GL14945KGBL |
45 kg | Fiber Drum |
Each for $7,642.71
|
|
||||
Description
Spectrum™ Chemical Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.Chemical Identifiers
| 31566-31-1 | |
| 358.56 | |
| 2,3-dihydroxypropyl octadecanoate |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO |
Specifications
| 31566-31-1 | |
| Poly Bottle | |
| 500 g | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO | |
| 358.56 |
| 100% | |
| C21H42O4 | |
| VBICKXHEKHSIBG-UHFFFAOYNA-N | |
| 2,3-dihydroxypropyl octadecanoate | |
| Food Grade |